Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010399.2 | NP_034529.1 | 230 | |||
GenBank mRNA | BC044208.1 | - | 241 | 5036676 | ||
M35248.1 | - | 230 |
Gene Symbol: H2-T10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161731.1 | - | 1 | 360 | ||
M35244.1 | - | 1 | 348 | |||
M35246.1 | - | 2 | 225 |
Gene Symbol: H2-T22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010397.4 | NP_034527.1 | 1 | 349 | ||
GenBank mRNA | AB359229.1 | - | 1 | 109 | ||
AK133985.1 | - | 2 | 281 | |||
AK161744.1 | - | 1 | 348 | |||
AK162874.1 | - | 2 | 251 | |||
BC003819.1 | - | 2 | 228 | 3484613 | ||
M35243.1 | - | 1 | 318 | |||
M35245.1 | - | 1 | 349 | |||
M35247.1 | - | 1 | 327 |