Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008209.4 | NP_032235.1 | 2 | 168 | ||
GenBank mRNA | AF010448.1 | - | 2 | 79 | ||
AF010452.1 | - | 2 | 79 | |||
AK041171.1 | - | 2 | 202 | |||
AK042119.1 | - | 2 | 194 | |||
AK137171.1 | - | 2 | 194 | |||
AK144040.1 | - | 2 | 193 | |||
AK162712.1 | - | 2 | 126 | |||
U94989.1 | - | 2 | 108 |