Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322434.1 | NP_001309363.1 | 3 | 396 | ||
NM_001322436.1 | NP_001309365.1 | 3 | 358 | |||
NM_001322437.1 | NP_001309366.1 | 3 | 461 | |||
NM_001322438.1 | NP_001309367.1 | 3 | 333 | |||
NM_001322439.1 | NP_001309368.1 | 3 | 396 | |||
NM_001322440.1 | NP_001309369.1 | 3 | 358 | |||
NM_001322441.1 | NP_001309370.1 | 3 | 461 | |||
NM_001322442.1 | NP_001309371.1 | 3 | 396 | |||
NM_001322443.1 | NP_001309372.1 | 3 | 377 | |||
NM_012207.2 | NP_036339.1 | 3 | 377 | |||
NM_021644.3 | NP_067676.2 | 3 | 377 | |||
XM_017016168.1 | XP_016871657.1 | 3 | 807 | |||
GenBank mRNA | AF132361.1 | - | 3 | 266 | ||
AF132362.1 | - | 1 | 278 | |||
AK091411.1 | - | 2 | 1743 | |||
AK223441.1 | - | 3 | 329 | |||
AK291547.1 | - | 3 | 329 | |||
AL117395.1 | - | 1 | 1093 | |||
BC039824.1 | - | 3 | 313 | 6061550 |