Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008214.4 | NP_032240.3 | 10 | 1286 | ||
GenBank mRNA | AK011685.1 | - | 10 | 1202 | ||
AK088824.1 | - | 10 | 1239 | |||
AK146922.1 | - | 10 | 1239 | |||
AK150081.1 | - | 10 | 1205 | |||
AK152329.1 | - | 10 | 1218 | |||
AK166826.1 | - | 10 | 1202 | |||
AK166892.1 | - | 10 | 1249 | |||
AK200496.1 | - | 1 | 20 | |||
BC020088.1 | - | 10 | 1221 | 4008479 | ||
U39473.1 | - | 10 | 1266 |