Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Has1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008215.2 | NP_032241.1 | 2 | 259 | ||
GenBank mRNA | AK053726.1 | - | 2 | 277 | ||
BC125424.1 | - | 2 | 238 | 40129939 | ||
BC125426.1 | - | 2 | 238 | 40129941 | ||
D82964.1 | - | 2 | 265 |