Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318186.1 | NP_001305115.1 | 10 | 852 | ||
NM_001318187.1 | NP_001305116.1 | 10 | 852 | |||
NM_001318188.1 | NP_001305117.1 | 11 | 924 | |||
NM_002140.4 | NP_002131.2 | 11 | 1085 | |||
NM_031262.3 | NP_112552.1 | 11 | 1085 | |||
NM_031263.3 | NP_112553.1 | 11 | 924 | |||
XM_005251960.2 | XP_005252017.1 | 11 | 903 | |||
XM_005251963.3 | XP_005252020.1 | 10 | 865 | |||
XM_005251965.2 | XP_005252022.1 | 10 | 823 | |||
XM_011518616.1 | XP_011516918.1 | 10 | 865 | |||
XM_017014668.1 | XP_016870157.1 | 11 | 903 | |||
XM_017014669.1 | XP_016870158.1 | 10 | 823 | |||
GenBank mRNA | AB209562.1 | - | 10 | 781 | ||
AB451263.1 | - | 9 | 698 | |||
AB451390.1 | - | 9 | 698 | |||
AK096385.1 | - | 10 | 710 | |||
AK123117.1 | - | 5 | 1650 | |||
AK291336.1 | - | 11 | 907 | |||
AK294067.1 | - | 10 | 882 | |||
AK300744.1 | - | 10 | 795 | |||
AK307962.1 | - | 11 | 815 | |||
AY911506.1 | - | 10 | 761 | |||
BC000355.2 | - | 10 | 820 | 2964383 | ||
BC014980.1 | - | 11 | 891 | 4906241 | ||
BC025321.1 | - | 11 | 843 | |||
CR456771.1 | - | 9 | 698 | |||
S74678.1 | - | 10 | 805 | |||
X72727.1 | - | 11 | 907 |