Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbb-bt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008220.5 | NP_032246.2 | 3 | 602 | ||
GenBank mRNA | AK002394.1 | - | 3 | 601 | ||
AK005496.1 | - | 3 | 601 | |||
AK011033.1 | - | 3 | 601 | |||
AK011049.1 | - | 616 | ||||
AK011050.1 | - | 3 | 602 | |||
AK011052.1 | - | 3 | 601 | |||
AK011057.1 | - | 3 | 601 | |||
AK011075.1 | - | 511 | ||||
AK012551.1 | - | 3 | 601 | |||
AK014364.1 | - | 3 | 602 | |||
AK028067.1 | - | 3 | 604 | |||
AK167615.1 | - | 3 | 601 | |||
AK168412.1 | - | 3 | 600 | |||
AK168477.1 | - | 3 | 601 | |||
AK168584.1 | - | 3 | 601 | |||
AK168819.1 | - | 3 | 601 |
Gene Symbol: Hbb-b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016956.3 | NP_058652.1 | 3 | 602 | ||
GenBank mRNA | BC027434.1 | - | 3 | 582 | 4920093 | |
BC032264.1 | - | 3 | 593 | 3988455 |