Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008224.4 | NP_032250.2 | 24 | 6299 | ||
GenBank mRNA | BC030898.1 | - | 8 | 1644 | ||
BC053742.1 | - | 24 | 6257 | 6509535 |