Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001005335.1 | NP_001005335.1 | 8 | 947 | ||
NM_001533.2 | NP_001524.2 | 8 | 1181 | |||
XM_011526886.1 | XP_011525188.1 | 8 | 970 | |||
XM_011526887.1 | XP_011525189.1 | 8 | 1014 | |||
XM_011526888.2 | XP_011525190.1 | 6 | 1069 | |||
XM_011526889.1 | XP_011525191.1 | 8 | 1065 | |||
GenBank mRNA | AB044547.1 | - | 8 | 1176 | ||
AK092828.1 | - | 2 | 1480 | |||
AK094966.1 | - | 1 | 2171 | |||
AK292115.1 | - | 8 | 946 | |||
AK301062.1 | - | 10 | 929 | |||
AK313652.1 | - | 8 | 1116 | |||
AY927544.1 | - | LIT3550 AY927544 |
1 | 20 | ||
BC069184.1 | - | 8 | 1120 | |||
X16135.1 | - | 8 | 1105 |