Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010408.3 | NP_034538.2 | 2 | 1177 | ||
GenBank mRNA | AF028737.1 | - | 2 | 797 | ||
AJ225123.1 | - | 2 | 1017 | |||
AK158322.1 | - | 2 | 1178 |