Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010411.2 | NP_034541.2 | 13 | 1118 | ||
GenBank mRNA | AF074881.1 | - | 13 | 1065 | ||
AF074882.1 | - | 7 | 492 | |||
AF098295.1 | - | 13 | 1043 | |||
AF125536.1 | - | 13 | 1050 | |||
AK037478.1 | - | 842 | ||||
AK145158.1 | - | 13 | 1084 | |||
BC139300.1 | - | 13 | 1146 | 8862322 | ||
BC139301.1 | - | 13 | 1146 | 8862323 | ||
JN651901.1 | - | 7 | 766 | |||
JN651902.1 | - | 6 | 724 | |||
JN651903.1 | - | 7 | 475 | |||
JN651904.1 | - | 4 | 320 |