Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 13 | 2212 | ||
NM_001284248.1 | NP_001271177.1 | 15 | 2724 | |||
NM_001284249.1 | NP_001271178.1 | 14 | 2321 | |||
NM_001284250.1 | NP_001271179.1 | 13 | 2209 | |||
NM_010412.3 | NP_034542.3 | 13 | 2209 | |||
GenBank mRNA | AF006602.2 | - | 12 | 1922 | ||
AF207748.1 | - | 12 | 1841 | |||
AK122325.2 | - | 8 | 1251 | |||
AK139202.1 | - | 14 | 2295 | |||
AK145972.1 | - | 13 | 2175 | |||
AK146483.1 | - | 13 | 2148 | |||
AK155335.1 | - | 13 | 2183 | |||
AK156008.1 | - | 7 | 3046 | |||
AK157159.1 | - | 7 | 3046 | |||
AK197372.1 | - | 1 | 184 | |||
AK213216.1 | - | 1 | 152 | |||
BC023309.1 | - | 4 | 762 | |||
BC049926.1 | - | 13 | 2145 | |||
BC058554.1 | - | 15 | 2676 | |||
BC060609.1 | - | 12 | 2003 | 5720438 |