Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004501.3 | NP_004492.2 | 8 | 1701 | ||
NM_031844.2 | NP_114032.2 | 8 | 1758 | |||
XM_017001115.1 | XP_016856604.1 | 8 | 1953 | |||
XM_017001116.1 | XP_016856605.1 | 8 | 1758 | |||
XM_017001117.1 | XP_016856606.1 | 8 | 1747 | |||
GenBank mRNA | AF068846.1 | - | 8 | 1754 | ||
AF461014.1 | - | 5 | 1279 | |||
AK126868.1 | - | 9 | 1499 | |||
AK130669.1 | - | 7 | 807 | |||
AK297119.1 | - | 9 | 1592 | |||
AK307661.1 | - | 8 | 1225 | |||
BC003367.1 | - | 8 | 1632 | 2900876 | ||
BC003621.2 | - | 8 | 1634 | 2966453 | ||
BC007950.2 | - | 8 | 1182 | |||
BC015782.1 | - | 8 | 1235 | 4857112 | ||
BC024767.1 | - | 8 | 1634 | 4993703 | ||
BC034925.1 | - | 5 | 625 | |||
X65488.1 | - | 8 | 1524 |