Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004501.3 | NP_004492.2 | 12, 13 | 2521 | ||
NM_031844.2 | NP_114032.2 | 12, 13 | 2578 | |||
XM_017001115.1 | XP_016856604.1 | 12, 13 | 2773 | |||
XM_017001116.1 | XP_016856605.1 | 12, 13 | 2578 | |||
XM_017001117.1 | XP_016856606.1 | 12, 13 | 2567 | |||
GenBank mRNA | AF068846.1 | - | 12, 13 | 2574 | ||
AF461014.1 | - | 9, 10 | 2099 | |||
AK126868.1 | - | 13, 14 | 2319 | |||
AK130669.1 | - | 11, 12 | 1627 | |||
AK297119.1 | - | 13, 14 | 2412 | |||
BC003367.1 | - | 12, 13 | 2452 | 2900876 | ||
BC003621.2 | - | 12, 13 | 2454 | 2966453 | ||
BC007950.2 | - | 12, 13 | 2002 | |||
BC015782.1 | - | 12, 13 | 2055 | 4857112 | ||
BC024767.1 | - | 12, 13 | 2454 | 4993703 | ||
BC034925.1 | - | 9, 10 | 1445 | |||
X65488.1 | - | 12, 13 | 2344 |