Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010418.2 | NP_034548.2 | 49 | 8027 | ||
GenBank mRNA | AF061529.1 | - | 49 | 7977 | ||
AF071173.1 | - | 49 | 7929 |