Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010421.4 | NP_034551.2 | 7 | 742 | ||
GenBank mRNA | AK075895.1 | - | 7 | 718 | ||
AK075911.1 | - | 7 | 743 | |||
AK080777.1 | - | 7 | 715 | |||
AK144168.1 | - | 7 | 870 | |||
AK150444.1 | - | 7 | 713 | |||
AK151960.1 | - | 7 | 717 | |||
AK159091.1 | - | 8 | 713 | |||
AK159814.1 | - | 7 | 728 | |||
AK159890.1 | - | 7 | 715 | |||
AK160112.1 | - | 7 | 702 | |||
AK167013.1 | - | 7 | 728 | |||
AK168186.1 | - | 7 | 742 | |||
AK170903.1 | - | 7 | 729 | |||
AK179213.1 | - | 2 | 89 | |||
AK218113.1 | - | 2 | 89 | |||
BC010755.1 | - | 7 | 711 | 4225233 | ||
U07631.1 | - | 7 | 734 | |||
X64331.1 | - | 7 | 685 |