Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010422.2 | NP_034552.1 | 3 | 494 | ||
GenBank mRNA | AK151149.1 | - | 3 | 431 | ||
AK159134.1 | - | 3 | 447 | |||
AK159178.1 | - | 3 | 468 | |||
AK160560.1 | - | 3 | 463 | |||
AK168825.1 | - | 3 | 468 | |||
U07633.1 | - | 3 | 466 | |||
Y00964.1 | - | 3 | 407 |