Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hey2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013904.1 | NP_038932.1 | 3 | 326 | ||
GenBank mRNA | AB093589.1 | - | 3 | 194 | ||
AF172287.1 | - | 3 | 326 | |||
AF173902.1 | - | 3 | 318 | |||
AF232240.1 | - | 3 | 322 | |||
AJ271867.1 | - | 3 | 326 | |||
AK031506.1 | - | 3 | 173 | |||
AK158000.1 | - | 3 | 327 | |||
BC103575.1 | - | 3 | 266 | 40045790 | ||
BC103576.1 | - | 3 | 269 | 40045791 | ||
BC105651.1 | - | 3 | 266 | 40046419 | ||
BC105652.1 | - | 3 | 269 | 40046420 |