Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008240.3 | NP_032266.3 | 3 | 850 | ||
GenBank mRNA | AK140847.1 | - | 3 | 787 | ||
BC082543.1 | - | 3 | 843 | 6808210 | ||
BC094569.1 | - | 2395 | ||||
L13204.1 | - | 2 | 697 |