Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008240.3 | NP_032266.3 | 3 | 2347 | ||
GenBank mRNA | AK140847.1 | - | 3 | 2284 | ||
BC082543.1 | - | 3 | 2340 | 6808210 | ||
BC094569.1 | - | 3892 | ||||
L13204.1 | - | 2 | 2195 |