Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DVL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004423.3 | NP_004414.3 | 8, 9 | 1037 | ||
XM_005247172.2 | XP_005247229.1 | 8, 9 | 1222 | |||
XM_011512513.2 | XP_011510815.1 | 5, 6 | 825 | |||
GenBank mRNA | AF006013.1 | - | 8, 9 | 908 | ||
AK093469.1 | - | 8, 9 | 1029 | |||
AK304686.1 | - | 9, 10 | 989 | |||
AL110278.1 | - | 4, 5 | 679 | |||
BC018289.1 | - | 6, 7 | 573 | |||
BC032459.1 | - | 8, 9 | 965 | 5200610 | ||
D86963.1 | - | 8, 9 | 1037 | |||
U48252.1 | - | 1, 2 | 142 | |||
U49262.1 | - | 8, 9 | 1018 | |||
U75651.1 | - | 8, 9 | 1290 |