Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016170.4 | NP_057254.1 | 3 | 1603 | ||
GenBank mRNA | AB593086.1 | - | 4 | 1290 | ||
BC006356.2 | - | 3 | 1588 | 4125397 | ||
BC006396.1 | - | 2 | 1860 | |||
BC018611.1 | - | 4 | 1199 |