Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008249.4 | NP_032275.2 | 5, 6 | 930 | ||
GenBank mRNA | AK090106.1 | - | 5, 6 | 901 | ||
AK146563.1 | - | 5, 6 | 931 | |||
AK166717.1 | - | 5, 6 | 917 | |||
AK167930.1 | - | 5, 6 | 882 | |||
AK168970.1 | - | 5, 6 | 909 | |||
AK169138.1 | - | 5, 6 | 909 | |||
AK181710.1 | - | 1, 2 | 24 | |||
AK206581.1 | - | 1, 2 | 24 | |||
AK212630.1 | - | 1, 2 | 24 | |||
AK214997.1 | - | 1, 2 | 24 | |||
BC138836.1 | - | 5, 6 | 875 | 8861858 | ||
BC138837.1 | - | 5, 6 | 875 | 8861859 | ||
M74555.1 | - | 5, 6 | 881 |