Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008250.2 | NP_032276.1 | 2 | 1082 | ||
GenBank mRNA | AK036435.1 | - | 2 | 1043 | ||
AK154090.1 | - | 2 | 760 | |||
BC050146.1 | - | 2 | 337 | |||
BC064068.1 | - | 2 | 1082 | 1380899 | ||
X58250.1 | - | 2 | 1198 |