Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110251.1 | NP_001103721.1 | 4 | 319 | ||
NM_013551.2 | NP_038579.2 | 4 | 377 | |||
GenBank mRNA | AK028234.1 | - | 4 | 244 | ||
AK143558.1 | - | 4 | 244 | |||
AK151020.1 | - | 4 | 343 | |||
AK153884.1 | - | 4 | 309 | |||
AK158160.1 | - | 1 | 1075 | |||
AK166734.1 | - | 4 | 275 | |||
AK167702.1 | - | 4 | 336 | |||
BB844509.1 | - | 4 | 320 | |||
BC003861.1 | - | 4 | 337 | 3487642 |