Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008251.3 | NP_032277.3 | 4, 5 | 255 | ||
GenBank mRNA | AK010763.1 | - | 261 | |||
BC083138.1 | - | 4, 5 | 213 | 6515849 | ||
BC088834.1 | - | 4, 5 | 197 | 6490162 | ||
BU923172.1 | - | 4, 5 | 196 | |||
X53476.1 | - | 4, 5 | 167 |