Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025427.3 | NP_001020598.1 | 3 | 360 | ||
NM_001039356.2 | NP_001034445.1 | 4 | 475 | |||
NM_001166535.1 | NP_001160007.1 | 3 | 398 | |||
NM_001166536.1 | NP_001160008.1 | 3 | 358 | |||
NM_001166537.1 | NP_001160009.1 | 3 | 652 | |||
NM_001166539.1 | NP_001160011.1 | 4 | 442 | |||
NM_001166540.1 | NP_001160012.1 | 3 | 365 | |||
NM_001166541.1 | NP_001160013.1 | 3 | 327 | |||
NM_001166542.1 | NP_001160014.1 | 3 | 325 | |||
NM_001166543.1 | NP_001160015.1 | 3 | 398 | |||
NM_001166544.1 | NP_001160016.1 | 3 | 360 | |||
NM_001166545.1 | NP_001160017.1 | 3 | 372 | |||
NM_001166546.1 | NP_001160018.1 | 3 | 360 | |||
NM_016660.3 | NP_057869.2 | 3 | 685 | |||
GenBank mRNA | AK010617.1 | - | 3 | 250 | ||
AK154957.1 | - | 3 | 281 | |||
AK155661.1 | - | 3 | 279 | |||
AK160036.1 | - | 3 | 249 | |||
AK169783.1 | - | 3 | 282 | |||
AK171334.1 | - | 3 | 314 | |||
AK182765.1 | - | 2 | 78 | |||
AK184734.1 | - | 2 | 78 | |||
AK185838.1 | - | 2 | 111 | |||
AK188296.1 | - | 2 | 111 | |||
AK199756.1 | - | 2 | 78 | |||
AK201985.1 | - | 2 | 111 | |||
BY165675.1 | - | 3 | 318 | |||
BY270784.1 | - | 3 | 361 | |||
CF730372.1 | - | 3 | 588 | |||
CF951929.1 | - | 3 | 251 | |||
GH454882.1 | - | 4 | 437 | |||
J04179.1 | - | 3 | 354 |
Gene Symbol: Hmga1-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166476.1 | NP_001159948.1 | 1 | 281 | ||
NM_001166477.1 | NP_001159949.1 | 2 | 248 | |||
GenBank mRNA | BC008125.1 | - | 2 | 233 | ||
BC013455.1 | - | 1 | 291 | 4219805 | ||
BC093496.1 | - | 1 | 248 | 3601855 |