Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010441.2 | NP_034571.1 | 4 | 1133 | ||
GenBank mRNA | AK028346.1 | - | 4 | 463 | ||
AK035164.1 | - | 1 | 3115 | |||
AK142059.1 | - | 4 | 495 | |||
BC052158.1 | - | 4 | 667 | 6506864 | ||
BC069985.1 | - | 4 | 361 | 6332337 | ||
BC085085.1 | - | 4 | 434 | 6334495 | ||
X58380.1 | - | 4 | 347 |