Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013552.2 | NP_038580.2 | 12, 13 | 1712 | ||
GenBank mRNA | AF031932.1 | - | 12, 13 | 1697 | ||
AF079222.1 | - | 13, 14 | 1661 | |||
AK143123.1 | - | 12, 13 | 1712 | |||
AK163410.1 | - | 12, 13 | 1592 | |||
BC021427.1 | - | 1836 | 5035341 | |||
X64550.1 | - | 8, 9 | 1325 |