Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013552.2 | NP_038580.2 | 8 | 761 | ||
GenBank mRNA | AF031932.1 | - | 8 | 746 | ||
AF079222.1 | - | 9 | 710 | |||
AK143123.1 | - | 8 | 761 | |||
AK161734.1 | - | 1 | 1226 | |||
AK163410.1 | - | 8 | 704 | |||
AK164410.1 | - | 8 | 735 | |||
BC021427.1 | - | 8 | 759 | 5035341 | ||
CN455388.1 | - | 8 | 763 | |||
X64550.1 | - | 4 | 374 |