Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008258.1 | NP_032284.1 | 3 | 329 | ||
GenBank mRNA | AK004909.1 | - | 3 | 344 | ||
AK010057.1 | - | 3 | 344 | |||
AK013122.1 | - | 3 | 341 | |||
AK014212.1 | - | 3 | 345 | |||
AK150410.1 | - | 3 | 348 | |||
AK152560.1 | - | 3 | 346 | |||
AK159784.1 | - | 3 | 343 | |||
AK167466.1 | - | 3 | 343 | |||
AK167961.1 | - | 3 | 333 | |||
AK168632.1 | - | 3 | 344 | |||
BC003758.1 | - | 3 | 318 | 3591830 | ||
U90123.1 | - | 3 | 329 |