Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008260.2 | NP_032286.1 | 2 | 632 | ||
GenBank mRNA | AK019022.1 | - | 3 | 514 | ||
X74938.1 | - | 2 | 629 |