Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170981.1 | NP_001164452.1 | 8 | 932 | ||
NM_001170982.1 | NP_001164453.1 | 7 | 906 | |||
NM_001170983.1 | NP_001164454.1 | 8 | 911 | |||
NM_001170984.1 | NP_001164455.1 | 8 | 908 | |||
NM_016884.3 | NP_058580.1 | 8 | 971 | |||
GenBank mRNA | AA797535.1 | - | 3 | 243 | ||
AF095257.1 | - | 8 | 925 | |||
AK010845.1 | - | 8 | 886 | |||
AK011336.1 | - | 8 | 904 | |||
AK012633.2 | - | 8 | 894 | |||
AK019958.1 | - | 8 | 2086 | |||
AK088245.1 | - | 8 | 948 | |||
AK088678.1 | - | 8 | 922 | |||
AK089061.1 | - | 8 | 912 | |||
AK150399.1 | - | 8 | 945 | |||
AK151446.1 | - | 8 | 948 | |||
AK153053.1 | - | 8 | 948 | |||
AK166590.1 | - | 8 | 889 | |||
AK168802.1 | - | 8 | 906 | |||
AK206075.1 | - | 3 | 277 | |||
AK217534.1 | - | 3 | 298 | |||
AK219753.1 | - | 2 | 103 | |||
BC004706.1 | - | 8 | 870 | 3499283 | ||
BC095922.1 | - | 7 | 847 | 4988965 | ||
CK620126.1 | - | 3 | 267 |