Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010461.2 | NP_034591.1 | 2 | 1590 | ||
GenBank mRNA | AB221615.1 | - | 2 | 529 | ||
BC139209.1 | - | 2 | 770 | 8862231 | ||
BC139212.1 | - | 3 | 710 | 8862234 | ||
X13721.1 | - | 2 | 1589 |