Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008273.2 | NP_032299.1 | 2 | 915 | ||
GenBank mRNA | AK054090.1 | - | 2 | 323 | ||
BC138115.1 | - | 2 | 915 | 8861134 |