Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 11 | 1299 | ||
NM_001122897.2 | NP_001116369.1 | 12 | 1536 | |||
NM_001285478.1 | NP_001272407.1 | 12 | 1556 | |||
NM_001285479.1 | NP_001272408.1 | 12 | 1494 | |||
NM_001285480.1 | NP_001272409.1 | 11 | 1260 | |||
NM_001285481.1 | NP_001272410.1 | 12 | 1458 | |||
NM_010470.3 | NP_034600.2 | 12 | 1497 | |||
GenBank mRNA | AK004938.1 | - | 12 | 1474 | ||
AK028181.1 | - | 12 | 1472 | |||
AK043260.1 | - | 11 | 1273 | |||
AK050234.1 | - | 12 | 1476 | |||
AK075725.1 | - | 11 | 1237 | |||
AK161041.1 | - | 11 | 1267 | |||
AK166161.1 | - | 12 | 1444 | |||
AK169740.1 | - | 12 | 1403 | |||
BC004053.1 | - | 8 | 777 | |||
BC020024.1 | - | 12 | 1447 | 3597151 |