Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 5 | 583 | ||
NM_001122897.2 | NP_001116369.1 | 6 | 820 | |||
NM_001285478.1 | NP_001272407.1 | 6 | 840 | |||
NM_001285479.1 | NP_001272408.1 | 6 | 781 | |||
NM_001285480.1 | NP_001272409.1 | 5 | 544 | |||
NM_001285481.1 | NP_001272410.1 | 6 | 742 | |||
NM_010470.3 | NP_034600.2 | 6 | 781 | |||
GenBank mRNA | AK004938.1 | - | 6 | 758 | ||
AK028181.1 | - | 6 | 759 | |||
AK043260.1 | - | 5 | 557 | |||
AK050234.1 | - | 6 | 760 | |||
AK075725.1 | - | 5 | 522 | |||
AK150784.1 | - | 6 | 826 | |||
AK161041.1 | - | 5 | 551 | |||
AK161084.1 | - | 5 | 3270 | |||
AK166161.1 | - | 6 | 728 | |||
AK169740.1 | - | 6 | 687 | |||
AK212728.1 | - | 2 | 142 | |||
BC004053.1 | - | 2 | 61 | |||
BC020024.1 | - | 6 | 731 | 3597151 | ||
BC107687.1 | - | 6 | 716 | |||
BU612149.1 | - | 5 | 584 | |||
X99642.1 | - | 4 | 424 |