Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 9 | 1123 | ||
NM_001122897.2 | NP_001116369.1 | 10 | 1360 | |||
NM_001285478.1 | NP_001272407.1 | 10 | 1380 | |||
NM_001285479.1 | NP_001272408.1 | 10 | 1318 | |||
NM_001285480.1 | NP_001272409.1 | 9 | 1084 | |||
NM_001285481.1 | NP_001272410.1 | 10 | 1282 | |||
NM_010470.3 | NP_034600.2 | 10 | 1321 | |||
GenBank mRNA | AK004938.1 | - | 10 | 1298 | ||
AK028181.1 | - | 10 | 1296 | |||
AK043260.1 | - | 9 | 1097 | |||
AK050234.1 | - | 10 | 1300 | |||
AK075725.1 | - | 9 | 1061 | |||
AK150784.1 | - | 10 | 1364 | |||
AK161041.1 | - | 9 | 1091 | |||
AK166161.1 | - | 10 | 1268 | |||
AK169740.1 | - | 10 | 1227 | |||
AK205238.1 | - | 2 | 154 | |||
BC004053.1 | - | 6 | 601 | |||
BC020024.1 | - | 10 | 1271 | 3597151 |