Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008277.2 | NP_032303.1 | 7 | 515 | ||
GenBank mRNA | AK149416.1 | - | 8 | 553 | ||
BC013343.1 | - | 7 | 539 | 4193670 | ||
D29987.1 | - | 8 | 553 | |||
X59530.1 | - | 7 | 505 |