Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006886.3 | NP_008817.1 | 2 | 247 | ||
GenBank mRNA | AF052955.1 | - | 2 | 226 | ||
AF077045.1 | - | 2 | 222 | |||
AK026556.1 | - | 2 | 219 | |||
AK315493.1 | - | 2 | 203 | |||
BC001690.1 | - | 2 | 197 | 2988582 | ||
BC003671.1 | - | 2 | 179 | 4052063 | ||
BC105811.1 | - | 2 | 241 | 6739745 | ||
BT007293.1 | - | 2 | 131 | |||
CB106920.1 | - | 2 | 262 | |||
DA202623.1 | - | 2 | 227 |
Gene Symbol: SLMO2-ATP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037929.1 | 7 | 835 | |||
NR_037930.1 | 5 | 576 | ||||
GenBank mRNA | CD522026.1 | - | 5 | 515 | ||
FY212904.1 | - | 7 | 724 |