Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169620.1 | - | 7 | 928 | ||
AK077456.1 | - | 7 | 985 | |||
AK140200.1 | - | 8 | 1073 | |||
AK144412.1 | - | 7 | 983 | |||
AK145829.1 | - | 7 | 985 | |||
AK146754.1 | - | 8 | 1046 | |||
AK146966.1 | - | 7 | 992 | |||
AK159186.1 | - | 7 | 991 | |||
AK188145.1 | - | 1 | 76 | |||
AK209624.1 | - | 1 | 76 | |||
BC088997.1 | - | 7 | 942 | |||
BC122563.1 | - | 6 | 916 | 40107557 | ||
BC125275.1 | - | 6 | 916 | 40107558 |