Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hrsp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008287.3 | NP_032313.2 | 2, 3 | 298 | ||
GenBank mRNA | AK005016.2 | - | 2, 3 | 274 | ||
BC092375.1 | - | 2, 3 | 232 | 5692223 | ||
BC125590.1 | - | 2, 3 | 198 | 40130105 | ||
BC125592.1 | - | 2, 3 | 198 | 40130107 | ||
BU559752.1 | - | 2, 3 | 301 |