Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010474.2 | NP_034604.1 | 2 | 1068 | ||
GenBank mRNA | AF019385.1 | - | 1069 | |||
AK087753.1 | - | 2 | 1047 | |||
AK132292.1 | - | 2 | 1079 | |||
BC009133.1 | - | 2 | 1030 | 3155049 |