Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012306.2 | NP_001012306.2 | 2, 3 | 274 | ||
GenBank mRNA | BC089581.1 | - | 2, 3 | 274 | 6768060 |