Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012306.2 | NP_001012306.2 | 3 | 930 | ||
NM_001161742.1 | NP_001155214.1 | 5 | 1209 | |||
NM_001161743.1 | NP_001155215.1 | 5 | 1156 | |||
NM_001161744.1 | NP_001155216.1 | 4 | 1148 | |||
NM_001161745.1 | NP_001155217.1 | 4 | 1095 | |||
GenBank mRNA | BC089581.1 | - | 3 | 930 | 6768060 | |
M77015.1 | - | 4 | 1021 |