Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013821.3 | NP_038849.2 | 3 | 483 | ||
GenBank mRNA | AB109387.1 | - | 2 | 159 | ||
AF031170.1 | - | 2 | 223 | |||
AK142267.1 | - | 3 | 485 | |||
AY046512.1 | - | 3 | 474 | |||
BC053501.1 | - | 3 | 469 | 30018625 |