Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013560.2 | NP_038588.2 | 3 | 662 | ||
GenBank mRNA | AK003119.1 | - | 3 | 583 | ||
AK010663.1 | - | 3 | 580 | |||
AK168927.1 | - | 3 | 580 | |||
AV458710.1 | - | 1 | 91 | |||
BC018257.1 | - | 3 | 602 | 3498180 | ||
BC090388.1 | - | 2 | 691 | |||
BC099463.1 | - | 3 | 557 | 30848584 | ||
U03560.1 | - | 3 | 519 | |||
U03561.1 | - | 4 | 486 | |||
U03562.1 | - | 4 | 420 | |||
X14686.1 | - | 3 | 564 | |||
X14687.1 | - | 3 | 609 |