Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLMO2-ATP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037929.1 | 7 | 776 | |||
NR_037930.1 | 5 | 517 | ||||
GenBank mRNA | CD522026.1 | - | 5 | 456 | ||
FY212904.1 | - | 7 | 665 |
Gene Symbol: ATP5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006886.3 | NP_008817.1 | 2 | 188 | ||
GenBank mRNA | AF052955.1 | - | 2 | 167 | ||
AF077045.1 | - | 2 | 163 | |||
AK026556.1 | - | 2 | 160 | |||
AK315493.1 | - | 2 | 144 | |||
BC001690.1 | - | 2 | 138 | 2988582 | ||
BC003671.1 | - | 2 | 120 | 4052063 | ||
BC105811.1 | - | 2 | 182 | 6739745 | ||
BT007293.1 | - | 2 | 72 | |||
CB106920.1 | - | 2 | 203 | |||
DA202623.1 | - | 2 | 168 |