Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010480.5 | NP_034610.1 | 2 | 225 | ||
GenBank mRNA | AK004658.1 | - | 2 | 227 | ||
AK077095.1 | - | 2 | 148 | |||
AK088975.1 | - | 2 | 141 | |||
AK146944.1 | - | 2 | 140 | |||
AK147641.1 | - | 2 | 99 | |||
AK157849.1 | - | 2 | 140 | |||
AK158753.1 | - | 1 | 174 | |||
AK161276.1 | - | 2 | 170 | |||
AK166696.1 | - | 2 | 140 | |||
AK166720.1 | - | 2 | 140 | |||
AK166729.1 | - | 2 | 131 | |||
AK166847.1 | - | 2 | 130 | |||
AK166867.1 | - | 2 | 139 | |||
AK166896.1 | - | 2 | 143 | |||
AK167008.1 | - | 2 | 140 | |||
AK167065.1 | - | 2 | 140 | |||
AK167086.1 | - | 2 | 139 | |||
AK167114.1 | - | 2 | 140 | |||
AK167171.1 | - | 2 | 139 | |||
AK167188.1 | - | 2 | 140 | |||
AK168407.1 | - | 2 | 140 | |||
AK168605.1 | - | 2 | 143 | |||
AK168620.1 | - | 2 | 140 | |||
AK183519.1 | - | 2 | 108 | |||
BC009003.1 | - | 2 | 110 | |||
BC018232.1 | - | 2 | 105 | |||
BC037151.1 | - | 2 | 121 | |||
BC046614.1 | - | 2 | 107 | 6491307 | ||
BC049124.1 | - | 2 | 220 | 6816250 | ||
BC057287.1 | - | 2 | 123 | |||
BC094024.1 | - | 2 | 122 | 4501952 | ||
M36830.1 | - | 1 | 66 |