Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008303.4 | NP_032329.1 | 3 | 503 | ||
GenBank mRNA | AK088121.1 | - | 3 | 334 | ||
AK133876.1 | - | 3 | 310 | |||
AK151935.1 | - | 3 | 313 | |||
AV570511.1 | - | 3 | 503 | |||
BC024385.1 | - | 3 | 262 | 5004427 | ||
BC099385.1 | - | 3 | 314 | 30848267 | ||
CJ238411.1 | - | 2 | 187 | |||
U09659.1 | - | 3 | 319 |